Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000545.6 | NP_000536.5 | 8 | 1768 | ||
NM_001306179.1 | NP_001293108.1 | 8 | 1768 | |||
XM_005253931.3 | XP_005253988.1 | 8 | 1821 | |||
GenBank mRNA | BC104908.1 | - | 8 | 1587 | 8143911 | |
BC104910.1 | - | 8 | 1587 | 8143913 | ||
BC143483.1 | - | 8 | 1588 | |||
HM116541.1 | - | 8 | 1710 | |||
HM116542.1 | - | 8 | 1384 | |||
HM116543.1 | - | 6 | 1173 | |||
HM116544.1 | - | 5 | 827 | |||
HM116545.1 | - | 5 | 697 | |||
HM116546.1 | - | 3 | 327 | |||
HM449088.1 | - | 8 | 1590 | |||
HM449089.1 | - | 7 | 1390 | |||
M57732.1 | - | 8 | 1590 |