Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005139.2 | NP_005130.1 | 6 | 646 | ||
GenBank mRNA | AK313945.1 | - | 6 | 421 | ||
BC000871.1 | - | 6 | 478 | 3460409 | ||
BX336217.2 | - | 6 | 646 | |||
CR407648.1 | - | 4 | 336 | |||
M20560.1 | - | 5 | 382 | |||
M63310.1 | - | 5 | 372 |