Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057132.3 | NP_476473.1 | 3 | 346 | ||
XM_006243699.1 | XP_006243761.1 | 5 | 614 | |||
XM_006243700.1 | XP_006243762.1 | 5 | 589 | |||
XM_006243701.1 | XP_006243763.1 | 5 | 709 | |||
GenBank mRNA | AY026068.1 | - | 3 | 346 | ||
AY026069.1 | - | 3 | 346 | |||
BC061732.1 | - | 5 | 523 | 5598621 | ||
D84477.1 | - | 3 | 316 |