Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057140.1 | NP_476481.1 | 13, 14 | 1667 | ||
GenBank mRNA | BC083859.1 | - | 13, 14 | 1667 | 7126254 | |
D14418.1 | - | 4, 5 | 425 |