Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057141.1 | NP_476482.1 | 5 | 301 | ||
XM_006253552.2 | XP_006253614.1 | 7 | 708 | |||
XM_006253553.2 | XP_006253615.1 | 6 | 404 | |||
XM_006253554.2 | XP_006253616.1 | 7 | 708 | |||
XM_006253555.2 | XP_006253617.1 | 7 | 708 | |||
GenBank mRNA | BC061867.1 | - | 6 | 388 | 5598872 | |
D17711.1 | - | 5 | 301 |