Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057142.1 | NP_476483.1 | 23 | 4353 | ||
GenBank mRNA | AF266164.1 | - | 4 | 571 | ||
EU348627.1 | - | 2 | 984 | |||
EU348628.1 | - | 4 | 1237 | |||
EU348633.1 | - | 3 | 398 | |||
EU348639.1 | - | 2 | 787 | |||
U66707.1 | - | 23 | 4353 |