Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057147.2 | NP_476488.2 | 2 | 185 | ||
XM_006248419.2 | XP_006248481.1 | 3 | 387 | |||
XM_006248420.2 | XP_006248482.1 | 2 | 422 | |||
XM_008768692.1 | XP_008766914.1 | 2 | 634 | |||
XM_008768693.1 | XP_008766915.1 | 2 | 636 | |||
GenBank mRNA | BC081746.1 | - | 2 | 185 | 7135589 |