Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057149.1 | NP_476490.1 | 1, 2 | 230 | ||
GenBank mRNA | AF187319.1 | - | 1, 2 | 230 |
Gene Symbol: LOC103689974 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770928.1 | XP_008769150.1 | 1, 2 | 480 |