Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760831.1 | XP_008759053.1 | 7 | 788 |
Gene Symbol: LOC103694886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773858.1 | XP_008772080.1 | 277 |