Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF3 SIRNA Assays ›
Gene Symbol:
TCF3
Gene Name:
transcription factor 3
Gene Aliases:
AGM8, E2A, E47, ITF1, TCF-3, VDIR, bHLHb21
Chromosome Location:
Chr.19: 1609290 - 1652590 on Build GRCh38
UniGene ID:
Hs.371282
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13876
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136139.2
NP_001129611.1
13, 14
1216
NM_003200.3
NP_003191.1
14, 15
1246
XM_006722852.3
XP_006722915.1
14, 15
1421
XM_006722853.3
XP_006722916.1
14, 15
1421
XM_006722855.3
XP_006722918.1
14, 15
1419
XM_006722856.3
XP_006722919.1
14, 15
1420
XM_006722857.3
XP_006722920.1
14, 15
1420
XM_006722858.3
XP_006722921.1
14, 15
1421
XM_011528215.2
XP_011526517.1
13, 14
2719
XM_011528216.2
XP_011526518.1
14, 15
1420
XM_011528217.2
XP_011526519.1
14, 15
1423
XM_011528220.2
XP_011526522.1
14, 15
1419
XM_011528221.2
XP_011526523.1
14, 15
1420
XM_011528225.2
XP_011526527.1
14, 15
1332
XM_011528227.2
XP_011526529.1
12, 13
1178
XM_017027177.1
XP_016882666.1
14, 15
1420
XM_017027178.1
XP_016882667.1
14, 15
1420
XM_017027180.1
XP_016882669.1
12, 13
1182
XM_017027181.1
XP_016882670.1
14, 15
1332
XM_017027182.1
XP_016882671.1
14, 15
1331
XM_017027184.1
XP_016882673.1
14, 15
1424
GenBank mRNA
BC011665.2
-
11, 12
990
BC110580.1
-
11, 12
1052
40007779
BG284440.1
-
2, 3
143
KF303591.1
-
11, 12
1024
M31222.1
-
1246
M31523.1
-
13, 14
1207
X52078.1
-
10, 11
973
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
B cell lineage commitment
transcription, DNA-templated
regulation of transcription, DNA-templated
B cell differentiation
immunoglobulin V(D)J recombination
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of muscle cell differentiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
transcription factor binding
mitogen-activated protein kinase kinase kinase binding
enhancer binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
repressing transcription factor binding
vitamin D response element binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0