Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057156.1 | NP_476497.1 | 2, 3 | 157 | ||
GenBank mRNA | L13445.1 | - | 2, 3 | 157 |