Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Decr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057197.2 | NP_476545.2 | 6 | 644 | ||
GenBank mRNA | BC059120.1 | - | 6 | 632 | 6920489 | |
CO385941.1 | - | 6 | 629 | |||
D00569.1 | - | 6 | 617 | |||
FQ209689.1 | - | 6 | 633 | |||
FQ210607.1 | - | 6 | 625 | |||
FQ210721.1 | - | 6 | 626 | |||
FQ212507.1 | - | 6 | 622 | |||
FQ218387.1 | - | 6 | 622 | |||
FQ219629.1 | - | 6 | 625 | |||
FQ219666.1 | - | 6 | 622 | |||
FQ220227.1 | - | 6 | 625 |