Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057205.2 | NP_476553.1 | 5 | 320 | ||
XM_006236936.2 | XP_006236998.1 | 4 | 303 | |||
GenBank mRNA | AF336829.1 | - | 5 | 329 | ||
BC081743.1 | - | 5 | 320 | 7124631 | ||
FQ234407.1 | - | 4 | 279 |