Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270860.1 | NP_001257789.1 | 11 | 1766 | ||
NM_001270861.1 | NP_001257790.1 | 11 | 1805 | |||
NM_058211.2 | NP_478118.1 | 11 | 1805 | |||
XM_008770481.1 | XP_008768703.1 | 10 | 1452 | |||
XM_008770482.1 | XP_008768704.1 | 10 | 1413 | |||
XM_008770483.1 | XP_008768705.1 | 10 | 1452 | |||
GenBank mRNA | AF069511.1 | - | 11 | 1766 | ||
AF070475.1 | - | 11 | 1805 | |||
AF080106.1 | - | 11 | 1805 |