Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SOX9 SIRNA Assays ›
Gene Symbol:
Sox9
Gene Name:
SRY (sex determining region Y)-box 9
Gene Aliases:
-
Chromosome Location:
Chr.10: 101288354 - 101294030 on Build Rnor_6.0
UniGene ID:
Rn.95086
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s138912
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sox9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_003750950.3
XP_003750998.1
1
206
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
cartilage condensation
ossification
branching involved in ureteric bud morphogenesis
cell fate specification
epithelial to mesenchymal transition
tissue homeostasis
positive regulation of protein phosphorylation
hair follicle development
positive regulation of mesenchymal cell proliferation
chondrocyte development
negative regulation of immune system process
heart valve development
heart valve morphogenesis
heart valve formation
endocardial cushion morphogenesis
chondrocyte differentiation involved in endochondral bone morphogenesis
chondrocyte hypertrophy
nucleosome assembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein complex assembly
protein phosphorylation
cytoskeleton organization
signal transduction
epidermal growth factor receptor signaling pathway
spermatogenesis
heart development
positive regulation of cell proliferation
negative regulation of cell proliferation
male gonad development
regulation of cell cycle process
positive regulation of gene expression
negative regulation of gene expression
positive regulation of epithelial cell migration
neural crest cell development
positive regulation of phosphatidylinositol 3-kinase signaling
single organismal cell-cell adhesion
male germ-line sex determination
cAMP-mediated signaling
cell differentiation
regulation of cell adhesion
extracellular matrix organization
male sex determination
negative regulation of ossification
negative regulation of bone mineralization
prostate gland development
negative regulation of epithelial cell differentiation
positive regulation of epithelial cell differentiation
mammary gland development
notochord development
otic vesicle formation
endocrine pancreas development
negative regulation of chondrocyte differentiation
positive regulation of chondrocyte differentiation
lacrimal gland development
protein localization to nucleus
somatic stem cell population maintenance
intrahepatic bile duct development
regulation of cell proliferation
regulation of apoptotic process
negative regulation of apoptotic process
protein kinase B signaling
cell fate commitment
regulation of cell differentiation
negative regulation of myoblast differentiation
positive regulation of protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of photoreceptor cell differentiation
oligodendrocyte differentiation
homeostasis of number of cells within a tissue
positive regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
cartilage development
Sertoli cell differentiation
Sertoli cell development
astrocyte fate commitment
retina development in camera-type eye
limb bud formation
retinal rod cell differentiation
endochondral bone morphogenesis
epithelial tube branching involved in lung morphogenesis
lung epithelial cell differentiation
prostate gland morphogenesis
epithelial cell proliferation involved in prostatic bud elongation
bronchus cartilage development
trachea cartilage development
intestinal epithelial structure maintenance
regulation of cell proliferation involved in tissue homeostasis
positive regulation of cartilage development
regulation of branching involved in lung morphogenesis
morphogenesis of a branching epithelium
lung smooth muscle development
negative regulation of biomineral tissue development
ERK1 and ERK2 cascade
Harderian gland development
cellular response to mechanical stimulus
cellular response to retinoic acid
cellular response to interleukin-1
cellular response to epidermal growth factor stimulus
cellular response to heparin
cellular response to transforming growth factor beta stimulus
renal vesicle induction
ureter development
ureter urothelium development
ureter smooth muscle cell differentiation
ureter morphogenesis
metanephric nephron tubule formation
negative regulation of canonical Wnt signaling pathway
cochlea morphogenesis
positive regulation of kidney development
positive regulation of branching involved in ureteric bud morphogenesis
positive regulation of extracellular matrix assembly
positive regulation of male gonad development
positive regulation of cell proliferation involved in heart morphogenesis
positive regulation of mesenchymal stem cell differentiation
regulation of epithelial cell proliferation involved in lung morphogenesis
negative regulation of mesenchymal cell apoptotic process
transcription regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein kinase activity
protein binding
beta-catenin binding
protein kinase A catalytic subunit binding
enhancer binding
bHLH transcription factor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
pre-mRNA intronic binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0