Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271267.1 | NP_001258196.1 | 2 | 261 | ||
XM_006237604.2 | XP_006237666.1 | 2 | 307 |