Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080581.1 | NP_542148.1 | 23 | 3266 | ||
GenBank mRNA | AB010467.1 | - | 23 | 3266 |