Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001153.4 | NP_001144.1 | 10 | 903 | ||
NM_001320698.1 | NP_001307627.1 | 12 | 2185 | |||
NM_001320700.1 | NP_001307629.1 | 10 | 837 | |||
NM_001320702.1 | NP_001307631.1 | 10 | 938 | |||
XM_017003943.1 | XP_016859432.1 | 13 | 2047 | |||
XM_017003944.1 | XP_016859433.1 | 13 | 2013 | |||
GenBank mRNA | AB209457.1 | - | 9 | 3379 | ||
AK058017.1 | - | 10 | 743 | |||
AK293177.1 | - | 10 | 810 | |||
AK293396.1 | - | 10 | 675 | |||
AK293406.1 | - | 10 | 691 | |||
AK303960.1 | - | 10 | 1462 | |||
AK315511.1 | - | 10 | 743 | |||
AY453398.1 | - | 9 | 687 | |||
BC000182.1 | - | 10 | 785 | 3352245 | ||
BC011659.1 | - | 10 | 763 | 4301675 | ||
BC063672.1 | - | 10 | 837 | 3908264 | ||
BX641114.1 | - | 1 | 3840 | |||
CR407681.1 | - | 8 | 655 | |||
D78152.1 | - | 9 | 717 | |||
FJ224287.1 | - | 10 | 728 | |||
M19383.1 | - | 10 | 728 | |||
M82809.1 | - | 10 | 728 |