Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080587.3 | NP_542154.3 | 9 | 1634 | ||
XM_008770135.1 | XP_008768357.1 | 8 | 2321 | |||
GenBank mRNA | S55933.1 | - | 9 | 1634 |