Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003206.3 | NP_003197.2 | 1 | 690 | ||
NM_198392.2 | NP_938206.1 | 1 | 691 | |||
GenBank mRNA | AF035718.1 | - | 1 | 674 | ||
AF047419.1 | - | 1 | 673 | |||
AL550251.3 | - | 1 | 691 | |||
BC025697.1 | - | 1 | 673 | 5221575 | ||
BT019660.1 | - | 1 | 414 | |||
CR450293.1 | - | 1 | 414 | |||
W73859.1 | - | 1 | 62 |