Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HEY2 SIRNA Assays ›
Gene Symbol:
Hey2
Gene Name:
hes-related family bHLH transcription factor with YRPW motif 2
Gene Aliases:
Herp1
Chromosome Location:
Chr.1: 29191170 - 29201257 on Build Rnor_6.0
UniGene ID:
Rn.58672
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s139176
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hey2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_130417.1
NP_569101.1
3
256
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013904, BC103575, AK158000, AF232240, AK031506, and others.
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
vasculogenesis
muscular septum morphogenesis
outflow tract morphogenesis
atrioventricular valve development
pulmonary valve morphogenesis
tricuspid valve morphogenesis
tricuspid valve formation
endocardial cushion to mesenchymal transition involved in heart valve formation
cardiac ventricle morphogenesis
cardiac left ventricle morphogenesis
cardiac right ventricle morphogenesis
ventricular trabecula myocardium morphogenesis
cardiac muscle hypertrophy
transcription from RNA polymerase II promoter
Notch signaling pathway
pattern specification process
heart development
anterior/posterior axis specification
positive regulation of heart rate
regulation of gene expression
negative regulation of gene expression
negative regulation of cardiac muscle cell apoptotic process
mesenchymal cell development
cardiac muscle hypertrophy in response to stress
ascending aorta morphogenesis
dorsal aorta morphogenesis
umbilical cord morphogenesis
cell fate commitment
regulation of auditory receptor cell differentiation
negative regulation of Notch signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
ventricular cardiac muscle cell development
positive regulation of cardiac muscle cell proliferation
cardiac epithelial to mesenchymal transition
heart trabecula formation
cardiac septum morphogenesis
ventricular septum morphogenesis
atrial septum morphogenesis
negative regulation of transcription initiation from RNA polymerase II promoter
labyrinthine layer blood vessel development
artery development
arterial endothelial cell differentiation
cardiac vascular smooth muscle cell development
coronary vasculature morphogenesis
pulmonary artery morphogenesis
protein-DNA complex assembly
cochlea development
vascular smooth muscle cell development
negative regulation of transcription regulatory region DNA binding
negative regulation of transcription from RNA polymerase II promoter involved in smooth muscle cell differentiation
regulation of vasculogenesis
transcription factor activity, RNA polymerase II core promoter sequence-specific
RNA polymerase II activating transcription factor binding
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor binding
microsatellite binding
histone deacetylase binding
sequence-specific DNA binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0