Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130813.1 | NP_570826.1 | 2 | 639 | ||
GenBank mRNA | AF294353.1 | - | 2 | 318 | ||
AF467250.1 | - | 2 | 639 |