Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093728.2 | NP_001087197.1 | 4 | 507 | ||
NM_001261817.1 | NP_001248746.1 | 4 | 492 | |||
NM_001261818.1 | NP_001248747.1 | 3 | 369 | |||
NM_001261819.1 | NP_001248748.1 | 3 | 354 | |||
NM_001261820.1 | NP_001248749.1 | 5 | 713 | |||
NM_001261821.1 | NP_001248750.1 | 5 | 698 | |||
NM_018679.5 | NP_061149.1 | 4 | 601 | |||
XM_005249339.2 | XP_005249396.1 | 3 | 262 | |||
XM_011514830.1 | XP_011513132.1 | 4 | 739 | |||
GenBank mRNA | AF260330.1 | - | 1 | 50 | ||
AF269223.1 | - | 3 | 285 | |||
AF536532.1 | - | 5 | 553 | |||
AF536533.1 | - | 4 | 540 | |||
AK057234.1 | - | 4 | 324 | |||
AK301778.1 | - | 4 | 383 | |||
AK301975.1 | - | 3 | 290 | |||
AK302022.1 | - | 3 | 255 | |||
AK302375.1 | - | 4 | 432 | |||
AK302456.1 | - | 4 | 413 | |||
AK315078.1 | - | 4 | 534 | |||
AK316403.1 | - | 4 | 316 | |||
AY069943.1 | - | 3 | 309 | |||
BC033729.1 | - | 4 | 453 | 5167179 | ||
BC040003.1 | - | 5 | 555 | 5741923 | ||
BC048418.1 | - | 4 | 531 | 5298037 | ||
BC090050.1 | - | 5 | 559 | 30387768 | ||
DC395417.1 | - | 4 | 440 |