Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130816.1 | NP_570829.1 | 2 | 217 | ||
XM_008767766.1 | XP_008765988.1 | 2 | 330 | |||
GenBank mRNA | AB079263.1 | - | 2 | 217 |