Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001059910.4 | XP_001059910.2 | 2 | 221 | ||
XM_002729144.4 | XP_002729190.1 | 2 | 835 |