Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006520.2 | NP_006511.1 | 4, 5 | 405 | ||
GenBank mRNA | AK311851.1 | - | 4, 5 | 340 | ||
BC000968.2 | - | 4, 5 | 333 | 3448623 | ||
CR450320.1 | - | 4, 5 | 279 | |||
CR542213.1 | - | 4, 5 | 279 | |||
DA342302.1 | - | 4, 5 | 396 | |||
DB492712.1 | - | 4, 5 | 403 | |||
U02556.1 | - | 4, 5 | 347 |