Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133311.1 | NP_579845.1 | 2 | 320 | ||
XM_008769421.1 | XP_008767643.1 | 2 | 472 | |||
GenBank mRNA | AF004774.2 | - | 2 | 269 | ||
AF269251.1 | - | 2 | 320 |