Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133312.1 | NP_579846.1 | 3 | 1104 | ||
XM_006241160.2 | XP_006241222.1 | 5 | 1566 | |||
XM_006241161.1 | XP_006241223.1 | 5 | 1565 | |||
XM_008765213.1 | XP_008763435.1 | 5 | 1571 | |||
GenBank mRNA | AF176023.1 | - | 3 | 1104 |