Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133316.1 | NP_579850.1 | 14 | 1301 | ||
XM_006238626.2 | XP_006238688.1 | 14 | 1329 | |||
XM_006238627.2 | XP_006238689.1 | 13 | 1226 | |||
XM_008763987.1 | XP_008762209.1 | 12 | 1287 | |||
GenBank mRNA | AF478683.1 | - | 14 | 1301 | ||
BC128728.1 | - | 12 | 1453 |