Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133321.2 | NP_579855.1 | 3 | 1228 | ||
XM_006248128.2 | XP_006248190.1 | 4 | 1317 | |||
XM_006248129.2 | XP_006248191.1 | 4 | 1454 | |||
XM_006248130.2 | XP_006248192.1 | 5 | 1616 | |||
XM_006248131.2 | XP_006248193.1 | 4 | 1457 | |||
XM_006248132.2 | XP_006248194.1 | 4 | 1408 | |||
XM_006248133.2 | XP_006248195.1 | 3 | 1192 | |||
XM_006248135.2 | XP_006248197.1 | 3 | 1649 | |||
XM_006248137.2 | XP_006248199.1 | 2 | 1470 | |||
XM_006248138.2 | XP_006248200.1 | 2 | 1109 | |||
XM_006248139.2 | XP_006248201.1 | 4 | 1465 | |||
XM_006248141.2 | XP_006248203.1 | 2 | 2339 | |||
XM_008768550.1 | XP_008766772.1 | 4 | 1425 | |||
XM_008768551.1 | XP_008766773.1 | 4 | 1379 | |||
XM_008768552.1 | XP_008766774.1 | 4 | 1336 | |||
XM_008768553.1 | XP_008766775.1 | 3 | 3592 | |||
GenBank mRNA | AY028455.1 | - | 3 | 1229 |