Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cetn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191842.1 | NP_001178771.1 | 4 | 371 | ||
GenBank mRNA | AF335277.2 | - | 3 | 261 | ||
FQ234873.1 | - | 4 | 304 |
Gene Symbol: LOC100912538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749163.3 | XP_003749211.1 | 4 | 1479 |