Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nap1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133402.2 | NP_596893.1 | 1 | 1867 | ||
GenBank mRNA | AB067678.1 | - | 1 | 1797 | ||
BC101932.1 | - | 1 | 1867 | 7938895 |