Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ITGA2 SIRNA Assays ›
Gene Symbol:
Itga2
Gene Name:
integrin, alpha 2
Gene Aliases:
CD49B
Chromosome Location:
Chr.2: 46996904 - 47097011 on Build Rnor_6.0
UniGene ID:
Rn.83597
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s139495
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Itga2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_345156.7
XP_345157.5
17
2352
Back To Top
More Information
Panther Classification:
Molecular Function -
integrin
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
positive regulation of leukocyte migration
movement of cell or subcellular component
substrate-dependent cell migration
hypotonic response
cell adhesion
cell-matrix adhesion
integrin-mediated signaling pathway
female pregnancy
cell proliferation
positive regulation of epithelial cell migration
positive regulation of alkaline phosphatase activity
response to organic cyclic compound
response to amine
response to muscle activity
positive regulation of smooth muscle cell migration
mammary gland development
positive regulation of cell projection organization
cell-substrate adhesion
positive regulation of collagen biosynthetic process
positive regulation of collagen binding
response to L-ascorbic acid
cell adhesion mediated by integrin
collagen-activated signaling pathway
wound healing
response to drug
positive regulation of DNA binding
skin morphogenesis
establishment of protein localization
positive regulation of translation
positive regulation of cell adhesion
positive regulation of smooth muscle contraction
focal adhesion assembly
mesodermal cell differentiation
positive regulation of smooth muscle cell proliferation
positive regulation of inflammatory response
positive regulation of positive chemotaxis
detection of mechanical stimulus involved in sensory perception of pain
positive regulation of transmission of nerve impulse
positive regulation of phagocytosis, engulfment
hepatocyte differentiation
response to parathyroid hormone
cellular response to mechanical stimulus
cellular response to estradiol stimulus
cellular response to organic cyclic compound
integrin binding
collagen binding
protein complex binding
collagen receptor activity
laminin binding
protein heterodimerization activity
cell adhesion molecule binding
collagen binding involved in cell-matrix adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0