Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133425.3 | NP_596916.1 | 2 | 942 | ||
GenBank mRNA | AF407168.3 | - | 2 | 942 | ||
BC086978.1 | - | 2 | 415 | 7191557 |