Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034830.1 | NP_001030002.1 | 11 | 1583 | ||
NM_001034831.1 | NP_001030003.1 | 10 | 1399 | |||
NM_133429.2 | NP_596920.2 | 11 | 1666 | |||
XM_006237326.2 | XP_006237388.1 | 9 | 1444 | |||
XM_006237327.2 | XP_006237389.1 | 12 | 1631 | |||
XM_006237328.2 | XP_006237390.1 | 9 | 1379 | |||
XM_006237329.2 | XP_006237391.1 | 9 | 1309 | |||
GenBank mRNA | AB019281.1 | - | 11 | 1583 | ||
AF216804.1 | - | 11 | 1666 | |||
AF216805.1 | - | 9 | 1385 | |||
AF216806.1 | - | 10 | 1399 | |||
AF216807.1 | - | 12 | 2330 |