Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133541.1 | NP_598225.1 | 7 | 1052 | ||
XM_006230205.2 | XP_006230267.1 | 7 | 1073 | |||
GenBank mRNA | L28801.1 | - | 7 | 1052 |