Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133542.2 | NP_598226.1 | 2 | 454 | ||
GenBank mRNA | AJ223184.1 | - | 2 | 454 |