Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003214.3 | NP_003205.2 | 10 | 975 | ||
GenBank mRNA | AF142482.1 | - | 10 | 929 | ||
AK226144.1 | - | 6 | 2335 | |||
BC010053.2 | - | 10 | 831 | |||
BC027877.1 | - | 8 | 713 | 5207501 | ||
BC091488.1 | - | 10 | 900 | 6187474 | ||
X94439.1 | - | 9 | 739 |