Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133558.1 | NP_598242.1 | 204 | |||
GenBank mRNA | AF163318.1 | - | 204 | |||
BC081733.1 | - | 254 | 7109438 |
Gene Symbol: LOC103690139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763102.1 | XP_008761324.1 | 2 | 3209 | ||
XM_008763103.1 | XP_008761325.1 | 3 | 1568 | |||
XM_008763104.1 | XP_008761326.1 | 2 | 576 | |||
XM_008763105.1 | XP_008761327.1 | 2 | 1410 | |||
XM_008763106.1 | XP_008761328.1 | 3 | 1488 | |||
XM_008763107.1 | XP_008761329.1 | 3 | 1115 |