Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004133.1 | NP_001004133.1 | 9 | 1739 | ||
NM_133440.1 | NP_597684.1 | 10 | 1866 | |||
NM_133441.1 | NP_597685.1 | 15 | 2360 | |||
NM_201350.1 | NP_958738.1 | 22 | 3179 | |||
XM_008760068.1 | XP_008758290.1 | 23 | 3120 | |||
GenBank mRNA | AF060116.1 | - | 9 | 1739 | ||
AJ131899.1 | - | 10 | 1866 | |||
AJ249562.1 | - | 15 | 2360 | |||
AY298755.1 | - | 22 | 3179 |
Gene Symbol: LOC103690160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230612.2 | XP_006230674.1 | 8 | 7308 | ||
XM_006230617.2 | XP_006230679.1 | 8 | 7281 | |||
XM_008760040.1 | XP_008758262.1 | 7 | 7287 | |||
XM_008760041.1 | XP_008758263.1 | 7 | 7287 | |||
XM_008760042.1 | XP_008758264.1 | 6 | 7278 | |||
XM_008760043.1 | XP_008758265.1 | 7 | 7260 | |||
XM_008760044.1 | XP_008758266.1 | 6 | 7239 |