Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004133.1 | NP_001004133.1 | 3 | 818 | ||
NM_133440.1 | NP_597684.1 | 3 | 924 | |||
NM_133441.1 | NP_597685.1 | 8 | 1418 | |||
NM_201350.1 | NP_958738.1 | 17 | 2267 | |||
XM_008760068.1 | XP_008758290.1 | 16 | 2178 | |||
GenBank mRNA | AF060116.1 | - | 3 | 818 | ||
AF141903.1 | - | 7 | 910 | |||
AF159048.1 | - | 7 | 928 | |||
AJ131899.1 | - | 3 | 924 | |||
AJ249562.1 | - | 8 | 1418 | |||
AY298755.1 | - | 17 | 2267 |
Gene Symbol: LOC103690160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230612.2 | XP_006230674.1 | 1 | 6366 | ||
XM_006230617.2 | XP_006230679.1 | 1 | 6366 | |||
XM_008760040.1 | XP_008758262.1 | 1 | 6366 | |||
XM_008760041.1 | XP_008758263.1 | 1 | 6366 | |||
XM_008760042.1 | XP_008758264.1 | 1 | 6366 | |||
XM_008760043.1 | XP_008758265.1 | 1 | 6366 | |||
XM_008760044.1 | XP_008758266.1 | 1 | 6366 |