Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133571.1 | NP_598255.1 | 12 | 1401 | ||
XM_006243702.2 | XP_006243764.1 | 12 | 1623 | |||
XM_006243703.1 | XP_006243765.1 | 13 | 1174 | |||
GenBank mRNA | AY724516.1 | - | 11 | 1422 | ||
D16236.1 | - | 12 | 1401 |