Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133584.1 | NP_598268.1 | 18, 19 | 2672 | ||
XM_006233260.2 | XP_006233322.1 | 18, 19 | 2600 | |||
GenBank mRNA | D89093.1 | - | 18, 19 | 2672 |