Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133586.2 | NP_598270.1 | 349 | |||
XM_008760588.1 | XP_008758810.1 | 4 | 318 | |||
XM_008760589.1 | XP_008758811.1 | 3 | 324 | |||
GenBank mRNA | AB010635.1 | - | 349 | |||
AB191005.1 | - | 386 |
Gene Symbol: LOC100910040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749155.3 | XP_003749203.1 | 3 | 414 |