Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133589.2 | NP_598273.2 | 3, 4 | 343 | ||
GenBank mRNA | BC078747.1 | - | 3, 4 | 343 | 7131278 | |
X77236.1 | - | 3, 4 | 308 |