Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001156.4 | NP_001147.1 | 10 | 1098 | ||
NM_001320879.1 | NP_001307808.1 | 9 | 1043 | |||
NM_001320880.1 | NP_001307809.1 | 10 | 1109 | |||
NM_004034.3 | NP_004025.1 | 11 | 1164 | |||
XM_017016162.1 | XP_016871651.1 | 11 | 1200 | |||
XM_017016163.1 | XP_016871652.1 | 10 | 1134 | |||
GenBank mRNA | AB062429.1 | - | 10 | 1056 | ||
AK222552.1 | - | 10 | 1066 | |||
AK295319.1 | - | 10 | 975 | |||
AK300086.1 | - | 8 | 865 | |||
AK301681.1 | - | 10 | 852 | |||
AK309141.1 | - | 10 | 860 | |||
AK312447.1 | - | 11 | 1137 | |||
AK313024.1 | - | 10 | 1041 | |||
BC001322.1 | - | 3 | 349 | |||
BC002632.2 | - | 10 | 1058 | 3610541 | ||
BQ689410.1 | - | 2 | 162 | |||
BT007187.1 | - | 9 | 1023 | |||
CR407686.1 | - | 9 | 1023 | |||
J04543.1 | - | 10 | 1083 |