Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000459.4 | NP_000450.2 | 10 | 1898 | ||
NM_001290077.1 | NP_001277006.1 | 9 | 1769 | |||
NM_001290078.1 | NP_001277007.1 | 8 | 1457 | |||
XM_005251561.1 | XP_005251618.1 | 10 | 1565 | |||
XM_005251563.1 | XP_005251620.1 | 9 | 1436 | |||
GenBank mRNA | AB086825.1 | - | 9 | 1425 | ||
AB208796.1 | - | 10 | 1576 | |||
AK291775.1 | - | 10 | 1586 | |||
AK294887.1 | - | 8 | 1458 | |||
AK295043.1 | - | 9 | 1495 | |||
AK308447.1 | - | 7 | 1009 | |||
BC035514.1 | - | 10 | 1573 | 5228999 | ||
EU826592.1 | - | 9 | 1319 | |||
L06139.1 | - | 10 | 1596 |