Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133609.2 | NP_598293.2 | 7 | 790 | ||
XM_006238630.2 | XP_006238692.1 | 7 | 761 | |||
GenBank mRNA | BC072507.1 | - | 7 | 790 | 7099973 |