Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133610.2 | NP_598294.1 | 10 | 2266 | ||
GenBank mRNA | AF185637.1 | - | 10 | 2225 | ||
AJ250280.1 | - | 10 | 2268 |