Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760700.1 | XP_008758922.1 | 4 | 342 | ||
GenBank mRNA | X83579.1 | - | 4 | 188 |